Learn More
2,3-Dimethoxycinnamic acid, predominantly trans, 98+%
Description
2,3-Dimethoxycinnamic acid is used in the synthesis of (2E,2E,2E)-N,N,N-(nitrilotri-2,1-ethanediyl)tris[3-(2,3-dimethoxyphenyl)-2-propenamide].
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
2,3-Dimethoxycinnamic acid is used in the synthesis of (2E,2′E,2′′E)-N,N′,N′′-(nitrilotri-2,1-ethanediyl)tris[3-(2,3-dimethoxyphenyl)-2-propenamide].
Notes
Keep container tightly sealed. Store in cool, dry conditions in well sealed containers. Incompatible with oxidizing agents.

Specifications
Specifications
Melting Point | 179°C to 183°C |
Quantity | 50 g |
Beilstein | 2695122 |
Formula Weight | 208.21 |
Percent Purity | ≥98% |
Chemical Name or Material | 2,3-Dimethoxycinnamic acid, predominantly trans |
Safety and Handling
Hazard Category | H315-H319-H335 |
Hazard Statement | GHS H Statement H315-H319-H335 Causes skin irritation. Causes serious eye irritation. May cause respiratory irritation. |
Precautionary Statement | P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c |
EINECSNumber | 231-249-5 |
TSCA | No |
Recommended Storage | Ambient temperatures |
RUO – Research Use Only
Your input is important to us. Please complete this form to provide feedback related to the content on this product.