Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

2,3-Dimethyl-1,3-butadiene, 98%, stabilized with BHT

Catalog No. ac163430050
Click to view available options
Quantity:
5 g
25 g
100 g
Packaging:
Glass bottle
This item is not returnable. View return policy
This item is not returnable. View return policy

CAS: 513-81-5 | C6H10 | 82.15 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 513-81-5
Molecular Formula C6H10
Molecular Weight (g/mol) 82.15
MDL Number MFCD00008595
InChI Key SDJHPPZKZZWAKF-UHFFFAOYSA-N
Synonym 2,3-dimethyl-1,3-butadiene, biisopropenyl, 2,3-dimethylbutadiene, diisopropenyl, 2,3-dimethylenebutane, 1,3-butadiene, 2,3-dimethyl, unii-61tuu25hco, 61tuu25hco, 2,3-dimethyl-buta-1,3-diene, 2,3-diene
PubChem CID 10566
IUPAC Name 2,3-dimethylbuta-1,3-diene
SMILES CC(=C)C(C)=C

Specifications

Melting Point -76.0°C
Color Colorless
Density 0.7260g/mL
Boiling Point 68.0°C to 69.0°C
Flash Point −20°C
Infrared Spectrum Authentic
Assay Percent Range 97.5% min. (GC)
Packaging Glass bottle
Refractive Index 1.438
Linear Formula H2C=C(CH3)C(CH3)=CH2
Quantity 5 g
Beilstein 01, III, 991
Fieser 01,276; 13,45
Specific Gravity 0.726
Merck Index 15, 3260
Stabilizer 100ppm BHT
Formula Weight 82.13
Percent Purity 98%
Physical Form Liquid
Chemical Name or Material 2, 3-Dimethyl-1, 3-butadiene, Stabilized with BHT
Show More Show Less
Hazard Category GHS Signal Word: Danger
Hazard Statement GHS H Statement
Highly flammable liquid and vapor.
Precautionary Statement GHS P Statement
Keep away from heat/sparks/open flames/hot surfaces.
- No smoking.
Ground/Bond container and receiving equipment.
EINECSNumber 208-172-

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.