Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

2,3-Dimethyl-2-butene, 1M solution in THF, AcroSeal™

Catalog No. ac438721000
Click to view available options
Quantity:
100 mL
Packaging:
AcroSeal™ Glass Bottle

CAS: 563-79-1 | C6H12 | 84.16 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 563-79-1, 109-99-9
Molecular Formula C6H12
Molecular Weight (g/mol) 84.16
InChI Key WGLLSSPDPJPLOR-UHFFFAOYSA-N
Synonym 2,3-dimethyl-2-butene, tetramethylethylene, 2-butene, 2,3-dimethyl, 2,3-dimethylbutene-2, 1,1,2,2-tetramethylethylene, tetramethylethene, 2,3-dimethyl-but-2-ene, ch3 2c=c ch3 2, 2-butene,3-dimethyl, acmc-1akps
PubChem CID 11250
IUPAC Name 2,3-dimethylbut-2-ene
SMILES CC(=C(C)C)C

Specifications

CAS Min % 9.33, 89.2
CAS Max % 10.8, 90.67
Color Colorless to Yellow
Density 0.8570g/mL
Flash Point −21°C
Packaging AcroSeal™ Glass Bottle
Linear Formula (CH3)2C=C(CH3)2
Quantity 100 mL
Specific Gravity 0.857
Formula Weight 84.16
Physical Form Liquid
Chemical Name or Material 2, 3-Dimethyl-2-butene
Show More Show Less
Hazard Category GHS Signal Word: Danger
Hazard Statement GHS H Statement
Highly flammable liquid and vapour.
Causes serious eye irritation.
May cause respiratory irritation.
May form explosive peroxides.
Suspected of causing cancer.
Precautionary Statement GHS P Statement
Keep away from heat/sparks/open flames/hot surfaces.
- No smoking.
IF ON SKIN (or hair): Take off immediately all contaminated clothing.
Rinse skin with water/shower.
IF SWALLOWED: rinse mouth.
Do NOT induce vomi
EINECSNumber 209-263-8

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.