Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

2,3-Dimethyl-4-nitrophenol 98.0+%, TCI America™
SDP

Catalog No. D260825G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
D260825G 25 g
D26085G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. D260825G Supplier TCI America Supplier No. D260825G
Add to Cart
Add to Cart

Chemical Identifiers

CAS 19499-93-5
Molecular Formula C8H9NO3
Molecular Weight (g/mol) 167.16
MDL Number MFCD01861284
InChI Key DAEZHJNJHQAEPE-UHFFFAOYSA-N
Synonym 4-Nitro-2,3-xylenol
PubChem CID 13331245
IUPAC Name 2,3-dimethyl-4-nitrophenol
SMILES CC1=C(O)C=CC(=C1C)[N+]([O-])=O

Specifications

Melting Point 125°C
Quantity 25 g
Formula Weight 167.16
Percent Purity ≥98.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material 2,3-Dimethyl-4-nitrophenol
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.