Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Alfa Aesar™ 2,3-Dimethylaniline, 98%

Manufacturer:  Alfa Aesar™ A1091836

Quantity 500g

 View more versions of this product

Catalog No. AAA1091836


  Check Availability
Add to cart

Specifications Description & Specifications

Chemical Identifiers

87-59-2
121.18
UN1711
14,10084
2,3-Dimethylaniline
C8H11N
MFCD00007732
742174
2,3-Xylidine

Specifications

96°C (204°F)
221° to 222°C
500g
C8H11N
MFCD00007732
742174
2,3-Xylidine
98%
Characteristic
2° to 3°C
87-59-2
121.18
UN1711
14,10084
2,3-Dimethylaniline

Safety and Handling

SDS & Safety Documents

GHS H Statement
H301-H311-H331-H373
Toxic if swallowed.
Toxic in contact with skin.
Toxic if inhaled.
May cause damage to organs through prolonged or repeated exposure.

GHS P Statement
P280h-P273-P309-P310
Wear protective gloves/clothing.
Avoid release to the environment.
IF exposed or if you feel unwell: Immediately call a POISON CENTER or doctor/physician.

Danger

DOTInformation : Transport Hazard Class: 6.1; Packing Group: II; Proper Shipping Name: XYLIDINES, LIQUID

RTECSNumber : ZE8750000

TSCA : TSCA

Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit