Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

2,3-Dimethylbenzaldehyde, 97%

Catalog No. AAH6442514
Click to view available options
Quantity:
5 g
25 g
100 g

CAS: 5779-93-1 | C9H10O | 134.178 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 5779-93-1
Molecular Formula C9H10O
Molecular Weight (g/mol) 134.178
MDL Number MFCD00798004
InChI Key UIFVCPMLQXKEEU-UHFFFAOYSA-N
Synonym benzaldehyde, dimethyl, hemellitaldehyde, dimethylbenzaldehyde, o-xylene-3-carboxaldehyde, dimethylformylbenzene, xylene-formaldehyde, pubchem3080, acmc-1ake8, 2,3-dimethyl-benzaldehyde, 2,3-dimethyl benzaldehyde
PubChem CID 34224
IUPAC Name 2,3-dimethylbenzaldehyde
SMILES CC1=CC=CC(=C1C)C=O

Specifications

Color Colorless to Yellow
Density 1.029 g/mL
Boiling Point 86°C to 88°C (10 mmHg)
Flash Point 101°C (214°F)
Refractive Index 1.553
Quantity 25 g
Beilstein 2238677
Formula Weight 134.18
Percent Purity 97%
Physical Form Liquid
Chemical Name or Material 2,3-Dimethylbenzaldehyde
Show More Show Less
TSCA No
Recommended Storage Ambient temperatures; Store under Nitrogen

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.