Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Alfa Aesar™ 2,3-Dimethylbenzaldehyde, 97%

Click to view available options
Quantity:
5g
25g
100g
5779-93-1
C9H10O
134.178
MFCD00798004
UIFVCPMLQXKEEU-UHFFFAOYSA-N
benzaldehyde, dimethyl,hemellitaldehyde,dimethylbenzaldehyde,o-xylene-3-carboxaldehyde,dimethylformylbenzene,xylene-formaldehyde,pubchem3080,acmc-1ake8,2,3-dimethyl-benzaldehyde,2,3-dimethyl benzaldehyde
34224
2,3-dimethylbenzaldehyde
CC1=CC=CC(=C1C)C=O
5779-93-1
C9H10O
134.178
MFCD00798004
UIFVCPMLQXKEEU-UHFFFAOYSA-N
benzaldehyde, dimethyl,hemellitaldehyde,dimethylbenzaldehyde,o-xylene-3-carboxaldehyde,dimethylformylbenzene,xylene-formaldehyde,pubchem3080,acmc-1ake8,2,3-dimethyl-benzaldehyde,2,3-dimethyl benzaldehyde
34224
2,3-dimethylbenzaldehyde
CC1=CC=CC(=C1C)C=O
Density 1.029g/mL
Refractive Index 1.5530
Boiling Point 86°C to 88°C (10mmHg)
Color Colorless to Yellow
Quantity 100g
Beilstein 2238677
Formula Weight 134.18
Physical Form Liquid
Percent Purity 97%
Chemical Name or Material 2,3-Dimethylbenzaldehyde