Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

2,3-Dimethylbenzene-1-carbonyl chloride, 97%, Thermo Scientific™

Catalog No. tl00220ea
Click to view available options
:
Amber Glass Bottle; 10g

Specifications

CAS 21900-46-9
CAS Min % 97.0
Assay Percent Range 97%
Molecular Formula C9H9ClO
Linear Formula (CH3)2C6H3C(O)Cl
Synonym 2,3-dimethylbenzene-1-carbonyl chloride, benzoyl chloride, dimethyl, xylic acid chloride, acmc-20aog3, 2,3 dimethylbenzoyl chloride, 2,3-dimethyl-benzoyl chloride, 2,3-dimethylbenzene-1-carbonylchloride, 2,3-dimethyl-1-benzenecarbonyl chloride, benzoyl chloride, 2,3-dimethyl-6ci,7ci,8ci,9ci
InChI Key WFNKMVDATNLZBX-UHFFFAOYSA-N
SMILES CC1=CC=CC(=C1C)C(=O)Cl
IUPAC Name 2,3-dimethylbenzoyl chloride
Molecular Weight (g/mol) 168.62
PubChem CID 2800899
Formula Weight 168.62
Chemical Name or Material 2, 3-Dimethylbenzene-1-carbonyl chloride
Show More Show Less
Hazard Category GHS Signal Word: Danger
Hazard Statement GHS H Statement:
Causes severe skin burns and eye damage.
Contact with water liberates toxic gas.
Reacts violently with water.
Precautionary Statement GHS P Statement:
IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
Wear eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Immediately call a POISON CENTER or doctor/physician.
Store in a dry place. Store in a closed container.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.