Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

2,3-Dimethylbenzoic acid, 98%

Catalog No. p-7080733
Click to view available options
Quantity:
5 g
25 g
100 g

CAS: 603-79-2 | C9H10O2 | 150.177 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 603-79-2
Molecular Formula C9H10O2
Molecular Weight (g/mol) 150.177
MDL Number MFCD00002479
InChI Key RIZUCYSQUWMQLX-UHFFFAOYSA-N
Synonym hemellitic acid, benzoic acid, 2,3-dimethyl, 2,3-dimethylbenzoicacid, 2,3-dimethyl benzoic acid, 2,3-dimethyl-benzoic acid, unii-7irp8ca267, 2,3-dimethylbenzenecarboxylic acid, vic-o-xylylic acid, methyl m-toluic acid, pubchem2549
PubChem CID 11782
ChEBI CHEBI:64823
IUPAC Name 2,3-dimethylbenzoic acid
SMILES CC1=CC=CC(=C1C)C(=O)O

Specifications

Melting Point 143°C to 147°C
Quantity 5 g
Odor Odorless
Beilstein 971590
Formula Weight 150.18
Percent Purity 98%
Chemical Name or Material 2,3-Dimethylbenzoic acid
Hazard Category H315-H319-H335
Hazard Statement GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
EINECSNumber 210-058-0
RTECSNumber DG8734000
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.