Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

2,3-Dimethylbutane, 98+%, ACROS Organics™

Click to view available options
Quantity:
5mL
100mL
1L
Packaging:
Glass bottle
79-29-8
C6H14
86.18
MFCD00008925
ZFFMLCVRJBZUDZ-UHFFFAOYSA-N
diisopropyl,biisopropyl,butane, 2,3-dimethyl,1,1,2,2-tetramethylethane,hsdb 76,unii-68isq7a432,ccris 6020,ch3 2chch ch3 2,acmc-1bmfq,dsstox_cid_5112
6589
2,3-dimethylbutane
CC(C)C(C)C
79-29-8
C6H14
86.18
MFCD00008925
ZFFMLCVRJBZUDZ-UHFFFAOYSA-N
diisopropyl,biisopropyl,butane, 2,3-dimethyl,1,1,2,2-tetramethylethane,hsdb 76,unii-68isq7a432,ccris 6020,ch3 2chch ch3 2,acmc-1bmfq,dsstox_cid_5112
6589
2,3-dimethylbutane
CC(C)C(C)C
Specific Gravity 0.662
Color Colorless
Packaging Glass bottle
Refractive Index 1.3740 to 1.3760
Density 0.6620g/mL
Melting Point -129.0°C
Boiling Point 58.0°C
Infrared Spectrum Authentic
Quantity 100mL
Flash Point −29°C
CAS Min % 98.0
CAS Max % 100.0
Assay Percent Range 98+%
Linear Formula (CH3)2CHCH(CH3)2
Beilstein 01,151
Solubility Information Solubility in water: insoluble in water. Other solubilities: soluble in most organic solvents
Formula Weight 86.18
Physical Form Liquid
Percent Purity ≥98%
Chemical Name or Material 2, 3-Dimethylbutane
Show More Show Less

GHS H Statement:
Causes skin irritation.
Toxic to aquatic life with long lasting effects.
May cause drowsiness or dizziness.
May be fatal if swallowed and enters airways.
Highly flammable liquid and vapor.

GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF SWALLOWED: Immediately call a POISON CENTRE or doctor/physician.
Do NOT induce vomiting.
Avoid release to the environment.
IF ON SKIN: Wash with plenty of soap and water.
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.
Ground/Bond container and receiving equipment.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS Signal Word: Danger