Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

2,3-Dimethylbutane 98.0+%, TCI America™
SDP

Catalog No. D069025ML Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
5 mL
25 mL
500 mL
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
D069025ML 25 mL
D06905ML 5 mL
D0690500ML 500 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. D069025ML Supplier TCI America Supplier No. D069025ML
Add to Cart
Add to Cart

Chemical Identifiers

CAS 79-29-8
Molecular Formula C6H14
Molecular Weight (g/mol) 86.178
MDL Number MFCD00008925
InChI Key ZFFMLCVRJBZUDZ-UHFFFAOYSA-N
Synonym diisopropyl, biisopropyl, butane, 2,3-dimethyl, 1,1,2,2-tetramethylethane, hsdb 76, unii-68isq7a432, ccris 6020, ch3 2chch ch3 2, acmc-1bmfq, dsstox_cid_5112
PubChem CID 6589
IUPAC Name 2,3-dimethylbutane
SMILES CC(C)C(C)C

Specifications

Color Colorless
Quantity 25 mL
Physical Form Clear Liquid at 20°C
UN Number 2457
Boiling Point 58°C
Formula Weight 86.18
Percent Purity ≥98.0% (GC)
Chemical Name or Material 2,3-Dimethylbutane
EINECSNumber (2)-0006
RTECSNumber EJ9350000
TSCA Yes
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.