Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

2,3-Dimethylindole 98.0+%, TCI America™
SDP

Catalog No. D1579250G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
25 g
250 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
D1579250G 250 g
D157925G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. D1579250G Supplier TCI America Supplier No. D1579250G
Add to Cart
Add to Cart

Chemical Identifiers

CAS 91-55-4
Molecular Formula C10H11N
Molecular Weight (g/mol) 145.205
MDL Number MFCD00005617
InChI Key PYFVEIDRTLBMHG-UHFFFAOYSA-N
Synonym 2,3-dimethylindole, 1h-indole, 2,3-dimethyl, indole, 2,3-dimethyl, 2,3-dimethyl indole, tfw7o9hwzk, unii-tfw7o9hwzk, 2,3-dimethyl-1h-indol, 2,3-dimethyl-indol, 2,3-dimethyl-indole, pubchem7334
PubChem CID 7053
IUPAC Name 2,3-dimethyl-1H-indole
SMILES CC1=C(NC2=CC=CC=C12)C

Specifications

Melting Point 107°C
Color Yellow
Boiling Point 285°C
Quantity 250 g
Formula Weight 145.21
Percent Purity ≥98.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material 2,3-Dimethylindole
RTECSNumber NL7185000
TSCA Yes
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.