Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

2,3-Dimethylmaleic anhydride, 97%

Catalog No. p-7046046
Click to view available options
Quantity:
5 g
25 g

CAS: 766-39-2 | C6H6O3 | 126.111 g/mol

2,3-Dimethylmaleic anhydride acts as a reagent in the preparation of maleimides. It is also employed as an amino group protecting agent for superoxide dismutase. Further, it serves as a reactant in the dissociation of ribosomal proteins. In addition to this, it is used in the preparation of dimethylmaleic acid dimethyl ester.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
2,3-Dimethylmaleic anhydride acts as a reagent in the preparation of maleimides. It is also employed as an amino group protecting agent for superoxide dismutase. Further, it serves as a reactant in the dissociation of ribosomal proteins. In addition to this, it is used in the preparation of dimethylmaleic acid dimethyl ester.

Solubility
Soluble in ethanol, ether and chloroform. Slightly soluble in water.

Notes
Moisture sensitive. Incompatible with strong oxidizing agents, strong acids and strong bases.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 766-39-2
Molecular Formula C6H6O3
Molecular Weight (g/mol) 126.111
MDL Number MFCD00005523
InChI Key MFGALGYVFGDXIX-UHFFFAOYSA-N
Synonym 2,3-dimethylmaleic anhydride, dimethylmaleic anhydride, 2,5-furandione, 3,4-dimethyl, pyrocinchonic anhydride, maleic anhydride, dimethyl, dimethylmaleic acid anhydride, 2,3-dimethylmaleicanhydride, unii-6pp3n541qa, 3,4-dimethyl-furan-2,5-dione, alpha,beta-dimethylmaleic anhydride
PubChem CID 13010
IUPAC Name 3,4-dimethylfuran-2,5-dione
SMILES CC1=C(C(=O)OC1=O)C

Specifications

Melting Point 93°C to 96°C
Boiling Point 222°C to 223°C
Quantity 5 g
Beilstein 112044
Sensitivity Moisture sensitive
Solubility Information Soluble in ethanol,ether and chloroform. Slightly soluble in water.
Formula Weight 126.11
Percent Purity 97%
Chemical Name or Material 2,3-Dimethylmaleic anhydride
Hazard Category H302-H315-H319-H335
Hazard Statement GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P270-P271-P280-P301+P312-P302+P352-P304+P340-P305+P351+P338-P312-P330-P332+P313-P362-P501c
EINECSNumber 212-165-8
RTECSNumber ON4025000
TSCA Yes
Recommended Storage Ambient temperatures; Store under Nitrogen

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.