Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

2,3-Dimethylmaleic Anhydride 97%, ACROS Organics™

Click to view available options
Quantity:
5g
50g
Packaging:
Glass bottle
766-39-2
C6H6O3
126.11
MFCD00005523
MFGALGYVFGDXIX-UHFFFAOYSA-N
2,3-dimethylmaleic anhydride,dimethylmaleic anhydride,2,5-furandione, 3,4-dimethyl,pyrocinchonic anhydride,maleic anhydride, dimethyl,dimethylmaleic acid anhydride,2,3-dimethylmaleicanhydride,unii-6pp3n541qa,3,4-dimethyl-furan-2,5-dione,alpha,beta-dimethylmaleic anhydride
13010
3,4-dimethylfuran-2,5-dione
CC1=C(C(=O)OC1=O)C
766-39-2
C6H6O3
126.11
MFCD00005523
MFGALGYVFGDXIX-UHFFFAOYSA-N
2,3-dimethylmaleic anhydride,dimethylmaleic anhydride,2,5-furandione, 3,4-dimethyl,pyrocinchonic anhydride,maleic anhydride, dimethyl,dimethylmaleic acid anhydride,2,3-dimethylmaleicanhydride,unii-6pp3n541qa,3,4-dimethyl-furan-2,5-dione,alpha,beta-dimethylmaleic anhydride
13010
3,4-dimethylfuran-2,5-dione
CC1=C(C(=O)OC1=O)C
Boiling Point 223.0°C
Melting Point 91.0°C to 96.0°C
Quantity 5g
Packaging Glass bottle
CAS Min % 96.0
CAS Max % 100.0
Assay Percent Range 97%
Beilstein 17, 445
Solubility Information Solubility in water: slightly soluble. Other solubilities: freely soluble in ethanol,ether and chloroform
Formula Weight 126.11
Percent Purity 97%
Chemical Name or Material 2, 3-Dimethylmaleic anhydride, 97%
Show More Show Less

GHS H Statement:
Causes serious eye damage.
Toxic in contact with skin.
Causes severe skin burns and eye damage.

GHS P Statement:
Wear protective gloves/protective clothing/eye protection/face protection.
Call a POISON CENTER or doctor/physician if you feel unwell.
IF ON SKIN: Gently wash with plenty of soap and water.
IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
Wear eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Immediately call a POISON CENTER or doctor/physician.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

MOISTURE SENSITIVE

EINECSNumber : 212-165-8

RTECSNumber : ON4025000

TSCA : TSCA