Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

2,3-Dimethylquinoxaline, 97%

Catalog No. AC116781000
Click to view available options
Quantity:
5 g
25 g
100 g

CAS: 2379-55-7 | C10H10N2 | 158.2 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 2379-55-7
MDL Number MFCD00006728
InChI Key FKHNZQFCDGOQGV-UHFFFAOYSA-N
PubChem CID 16925
IUPAC Name 2,3-dimethylquinoxaline
SMILES CC1=NC2=CC=CC=C2N=C1C

Specifications

CAS Min % 96.0
CAS Max % 100.0
Melting Point 102°C to 107°C
Color Beige
Infrared Spectrum Authentic
Assay Percent Range 97%
Molecular Formula C10H10N2
Quantity 100 g
Beilstein 23, 191
Synonym quinoxaline, 2,3-dimethyl, aurora ka-3449, quinoxaline,3-dimethyl, 2,3-dimethyl-quinoxalin, 2,3-dimethyl-quinoxaline, acmc-1cg5s, quinoxaline,2,3-dimethyl, salor-int l498777-1ea, 5-23-07-00267 beilstein handbook reference, chembl67865
Molecular Weight (g/mol) 158.204
Formula Weight 158.2
Percent Purity ≥96.0%
Physical Form Crystalline Powder
Chemical Name or Material 2, 3-Dimethylquinoxaline, 97%
Show More Show Less
EINECSNumber 219-162-0
RTECSNumber VD1960000
TSCA TSCA

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.