Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

2,3-Dimethylthiophene, 97%, ACROS Organics™

Click to view available options
Quantity:
1g
5g
25g
632-16-6
C6H8S
112.2
BZYUMXXOAYSFOW-UHFFFAOYSA-N
thiophene, 2,3-dimethyl,2,3-dimethyl-thiophene,2,3,dimethylthiophene,pubchem7751,2.3-dimethylthiophene,dimethylthiophene,2,3-dirnethylthiophene
34295
2,3-dimethylthiophene
CC1=C(SC=C1)C
632-16-6
C6H8S
112.2
BZYUMXXOAYSFOW-UHFFFAOYSA-N
thiophene, 2,3-dimethyl,2,3-dimethyl-thiophene,2,3,dimethylthiophene,pubchem7751,2.3-dimethylthiophene,dimethylthiophene,2,3-dirnethylthiophene
34295
2,3-dimethylthiophene
CC1=C(SC=C1)C
Infrared Spectrum Authentic
Color Colorless to Yellow
Quantity 1g
Packaging Glass bottle
Refractive Index 1.5170 to 1.5210
Flash Point >1°C
Assay Percent Range 96% min. (GC)
Formula Weight 112.2
Physical Form Liquid
Percent Purity 97%
Chemical Name or Material 2, 3-Dimethylthiophene
Show More Show Less

GHS Signal Word: Danger

EINECSNumber : 211-170-2