Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

2,3-Diphenyl-2-cyclopropen-1-one, 99% (dry wt.), may cont up to 5% water

Catalog No. p-7026131
Click to view available options
Quantity:
1 g
5 g
25 g

CAS: 886-38-4 | C15H10O | 206.24 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 886-38-4
Molecular Formula C15H10O
Molecular Weight (g/mol) 206.24
MDL Number MFCD00001311
InChI Key HCIBTBXNLVOFER-UHFFFAOYSA-N
Synonym diphenylcyclopropenone, diphencyprone, 2,3-diphenylcyclopropenone, 2,3-diphenylcycloprop-2-enone, cyclopropenone, diphenyl, dpcp, 2-cyclopropen-1-one, 2,3-diphenyl, 1,2-diphenylcyclopropen-3-one, cyclopropenone, 2,3-diphenyl, unii-i7g14nw5ec
PubChem CID 65057
ChEBI CHEBI:53074
IUPAC Name 2,3-diphenylcycloprop-2-en-1-one
SMILES O=C1C(=C1C1=CC=CC=C1)C1=CC=CC=C1

Specifications

Melting Point 119°C to 121°C
Quantity 1 g
Beilstein 608049
Merck Index 14,3308
Formula Weight 206.24
Percent Purity 99%
Assay (dry wt.), may cont up to 5% water
Chemical Name or Material 2,3-Diphenyl-2-cyclopropen-1-one
Hazard Category H315-H317-H319-H335
Hazard Statement GHS H Statement
H317
May cause an allergic skin reaction.
Precautionary Statement P261-P264b-P271-P272-P280-P302+P352-P304+P340-P305+P351+P338-P312-P333+P313-P362-P501c
EINECSNumber 212-948-4
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.