Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
2,3-Lutidine, 99%
Click to view available options
Quantity:
25 g
100 g
500 g
Description
2,3-Lutidine is a lansoprazole intermediates. It is used as pharmaceutical intermediate.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
2,3-Lutidine is a lansoprazole intermediates. It is used as pharmaceutical intermediate.
Solubility
Soluble in water 95 gl/ @ 20˚C.
Notes
Store away from strong oxidizing agents. Keep container tightly closed. Store in cool, dry conditions in well sealed containers.
2,3-Lutidine is a lansoprazole intermediates. It is used as pharmaceutical intermediate.
Solubility
Soluble in water 95 gl/ @ 20˚C.
Notes
Store away from strong oxidizing agents. Keep container tightly closed. Store in cool, dry conditions in well sealed containers.

Specifications
Specifications
Melting Point | -17°C |
Density | 0.948 |
Boiling Point | 160°C to 161°C |
Flash Point | 50°C (122°F) |
Odor | Unpleasant |
Refractive Index | 1.508 |
Quantity | 100 g |
UN Number | UN2929 |
Beilstein | 106418 |
Solubility Information | Soluble in water 95 gl/ @ 20?C. |
Show More |
Safety and Handling
Hazard Category | H226-H302+H312+H332-H315-H319-H335 |
Hazard Statement | GHS H Statement H331-H226-H302-H312-H315-H319-H335 Toxic if inhaled. Flammable liquid and vapor. Harmful if swallowed. Harmful in contact with skin. Causes skin irritation. Causes serious eye irritation. May cause respiratory irritation. |
Precautionary Statement | P210-P233-P235-P240-P241-P242-P243-P261-P264b-P270-P271-P280-P301+P312-P303+P361+P353-P304+P340-P305+P351+P338-P312-P330-P332+P313-P363-P370+P378q-P501c |
DOTInformation | Hazard Class: 3; Packaging Group: III |
EINECSNumber | 209-514-1 |
TSCA | Yes |
Recommended Storage | Ambient temperatures |
RUO – Research Use Only
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction