Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

2-(3-Methoxyphenyl)cyclohexanone, 90%, Tech., Thermo Scientific™

Catalog No. AC172860050
Click to view available options
Quantity:
5 g
Packaging:
Glass Bottle

This Thermo Scientific brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 15547-89-4
Molecular Formula C13H16O2
Molecular Weight (g/mol) 204.269
MDL Number MFCD00001630
InChI Key KACQSVYTBQDRGP-UHFFFAOYSA-N
Synonym 2-3-methoxyphenyl cyclohexanone, 2-3-methoxyphenyl cyclohexan-1-one, cyclohexanone, 2-3-methoxyphenyl, acmc-20aoq7, 2-3-methoxyphenyl-cyclohexanone, 2-3-methoxy-phenyl-cyclohexanone, cyclohexanone,2-3-methoxyphenyl, 2-3-methoxyphenyl-cyclohexan-1-one, 2-3-methoxyphenyl cyclohexanone, technical grade
PubChem CID 85868
IUPAC Name 2-(3-methoxyphenyl)cyclohexan-1-one
SMILES COC1=CC=CC(=C1)C2CCCCC2=O

Specifications

CAS Min % 88.0
CAS Max % 100.0
Color Colorless
Density 1.1000g/mL
Infrared Spectrum Authentic
Assay Percent Range 90%
Packaging Glass Bottle
Linear Formula CH3OC6H4C6H9(=O)
Refractive Index 1.5450 to 1.5490 (20°C, 589nm)
Quantity 5 g
Formula Weight 204.27
Percent Purity ≥88%
Grade Technical
Physical Form Liquid
Chemical Name or Material 2-(3-Methoxyphenyl)cyclohexanone, tech. 90%
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.