Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

2,3-Naphthalenedicarboxylic Anhydride 95.0+%, TCI America™
SDP

Catalog No. N0530250G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
25 g
250 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
N0530250G 250 g
N053025G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. N0530250G Supplier TCI America Supplier No. N0530250G
Add to Cart
Add to Cart

Chemical Identifiers

CAS 716-39-2
Molecular Formula C12H6O3
Molecular Weight (g/mol) 198.177
MDL Number MFCD00059091
InChI Key IZJDCINIYIMFGX-UHFFFAOYSA-N
PubChem CID 69743
IUPAC Name benzo[f][2]benzofuran-1,3-dione
SMILES C1=CC=C2C=C3C(=CC2=C1)C(=O)OC3=O

Specifications

Melting Point 248°C
Boiling Point 380°C
Quantity 250 g
Formula Weight 198.18
Percent Purity ≥95.0% (GC,T)
Physical Form Crystalline Powder
Chemical Name or Material 2,3-Naphthalenedicarboxylic Anhydride
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.