Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

2,3-Pyridinedicarboxylic acid, 99%

Catalog No. AC131850250
Click to view available options
Quantity:
25 g
100 g
500 g
Packaging:
Plastic bottle

CAS: 89-00-9 | C7H5NO4 | 167.12 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 89-00-9
Molecular Formula C7H5NO4
Molecular Weight (g/mol) 167.12
MDL Number MFCD00006295
InChI Key GJAWHXHKYYXBSV-UHFFFAOYSA-N
Synonym quinolinic acid, 2,3-pyridinedicarboxylic acid, quinolinate, pyridine-2,3-dicarboxylate, unii-f6f0hk1urn, 2,3-pyridinedicarboxylicacid, pyridine-2,3-carboxylate, f6f0hk1urn, pyridin-2,3-dicarbonsaeure, 2,3-pyridinedicarboxylate
PubChem CID 1066
ChEBI CHEBI:16675
IUPAC Name pyridine-2,3-dicarboxylic acid
SMILES C1=CC(=C(N=C1)C(=O)O)C(=O)O

Specifications

CAS Min % 98.5
CAS Max % 100.0
Melting Point 188°C to 190°C
Infrared Spectrum Authentic
Assay Percent Range 99%
Packaging Plastic bottle
Quantity 25 g
Beilstein 22, 150
Merck Index 15, 8190
Solubility Information Solubility in water: 0.55%. Other solubilities: soluble in alkalies,slightly soluble in alcohol,practically insoluble in ether and benzene
Formula Weight 167.12
Percent Purity 99%
Physical Form Crystalline Powder
Chemical Name or Material 2, 3-Pyridinedicarboxylic acid, 99%
Show More Show Less
Hazard Category GHS Signal Word: Warning
Hazard Statement GHS H Statement
May cause respiratory irritation.
Causes skin irritation.
Causes serious eye irritation.
Precautionary Statement GHS P Statement
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses,if present and easy to do.
Continue rinsing.
EINECSNumber 201-874-8
RTECSNumber US7967250
TSCA TSCA

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.