Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

2,3-Pyridinedicarboxylic Anhydride 95.0+%, TCI America™
SDP

Catalog No. P122225G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
P122225G 25 g
P12225G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. P122225G Supplier TCI America Supplier No. P122225G
Add to Cart
Add to Cart

Chemical Identifiers

CAS 699-98-9
Molecular Formula C7H3NO3
Molecular Weight (g/mol) 149.105
MDL Number MFCD00005915
InChI Key MCQOWYALZVKMAR-UHFFFAOYSA-N
Synonym 2,3-pyridinedicarboxylic anhydride, furo 3,4-b pyridine-5,7-dione, quinolinic anhydride, 2,3-pyridinedicarboxylicanhydride, pyridine-2,3-dicarboxylic anhydride, 5h,7h-furo 3,4-b pyridine-5,7-dione, furano 3,4-b pyridine-5,7-dione, pyridine-2,3-dicarboxylic acid anhydride, 2,3-pyridinedicarboxylic anhydride quinolinic anhydride
PubChem CID 69688
IUPAC Name furo[3,4-b]pyridine-5,7-dione
SMILES C1=CC2=C(C(=O)OC2=O)N=C1

Specifications

Melting Point 138°C
Color White-Yellow
Quantity 25 g
Formula Weight 149.11
Percent Purity ≥95.0% (T)
Physical Form Crystalline Powder
Chemical Name or Material 2,3-Pyridinedicarboxylic Anhydride
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.