Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
2(3)-tert-Butyl-4-methoxyphenol, 96%
Click to view available options
Quantity:
50 g
250 g
1000 g
Description
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Suitable for battery materials development.
Applications
2(3)-tert-Butyl-4-methoxyphenol is used as an industrial antioxidant. It is used as a food additive, preservative in food packaging, animal feed, cosmetics, rubber, and petroleum products.
Solubility
Soluble in ethanol, methanol, chloroform, fats, and oils. Insoluble in water.
Notes
Store in the dark place. Store away from light, oxidizing agents.
2(3)-tert-Butyl-4-methoxyphenol is used as an industrial antioxidant. It is used as a food additive, preservative in food packaging, animal feed, cosmetics, rubber, and petroleum products.
Solubility
Soluble in ethanol, methanol, chloroform, fats, and oils. Insoluble in water.
Notes
Store in the dark place. Store away from light, oxidizing agents.

Specifications
Specifications
Melting Point | 55°C to 65°C |
Quantity | 1000 g |
Sensitivity | Light sensitive |
Merck Index | 14,1547 |
Solubility Information | Soluble in ethanol,methanol,chloroform,fats,and oils. Insoluble in water. |
Formula Weight | 180.25 |
Percent Purity | 96% |
Chemical Name or Material | 2(3)-tert-Butyl-4-methoxyphenol |
Safety and Handling
Hazard Category | H302-H315-H319-H335-H350 |
Hazard Statement | GHS H Statement H351-H302 Suspected of causing cancer. Harmful if swallowed. |
Precautionary Statement | P201-P202-P261-P264b-P270-P271-P280i-P281-P301+P312-P302+P352-P304+P340-P305+P351+P338-P308+P313-P330-P332+P313-P362-P501c |
EINECSNumber | 246-563-8 |
TSCA | Yes |
Recommended Storage | Ambient temperatures |
RUO – Research Use Only
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction