Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

2,4,5-Tribromoimidazole, 97%, Thermo Scientific™

Catalog No. p-3779692
Click to view available options
Quantity:
100 g
Packaging:
Glass bottle

This Thermo Scientific brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 2034-22-2
MDL Number MFCD00005184
InChI Key JCGGPCDDFXIVQB-UHFFFAOYSA-N
Synonym 2,4,5-tribromoimidazole, 1h-imidazole, 2,4,5-tribromo, imidazole, 2,4,5-tribromo, 2,4,5-tribromoimidazo, tribromoimidazole, pubchem7150, 2,5-tribromoimidazole, acmc-1cmbx, timtec-bb sbb001252, imidazole,4,5-tribromo
PubChem CID 16253
IUPAC Name 2,4,5-tribromo-1H-imidazole
SMILES C1(=C(N=C(N1)Br)Br)Br

Specifications

CAS Min % 96.0
CAS Max % 100.0
Color Beige-Yellow
Infrared Spectrum Authentic
Assay Percent Range 97%
Packaging Glass bottle
Molecular Formula C3HBr3N2
Quantity 100 g
Beilstein 23, 49
Molecular Weight (g/mol) 304.767
Formula Weight 304.76
Percent Purity ≥96.0%
Physical Form Powder
Chemical Name or Material 2, 4, 5-Tribromoimidazole
Show More Show Less
Hazard Category GHS Signal Word: Danger
Hazard Statement GHS H Statement:
Toxic if swallowed.
Precautionary Statement GHS P Statement:
IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.