Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

2,4,5-Trichlorobenzenesulfonic Acid Hydrate 98.0+%, TCI America™
SDP

Catalog No. B024825G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
B024825G 25 g
B02485G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. B024825G Supplier TCI America Supplier No. B024825G
Add to Cart
Add to Cart

Chemical Identifiers

CAS 6378-25-2
Molecular Formula C6H3Cl3O3S
Molecular Weight (g/mol) 261.497
MDL Number MFCD00070539
InChI Key LEDKKDPOPIKMSZ-UHFFFAOYSA-N
PubChem CID 80786
IUPAC Name 2,4,5-trichlorobenzenesulfonic acid
SMILES C1=C(C(=CC(=C1Cl)Cl)Cl)S(=O)(=O)O

Specifications

Color White
Quantity 25 g
UN Number 2585
Formula Weight 261.50
Percent Purity ≥98.0% (T)
Physical Form Crystalline Powder
Chemical Name or Material 2,4,5-Trichlorobenzenesulfonic Acid Hydrate
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.