Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

2,4,5-Trifluoronitrobenzene 98.0+%, TCI America™
SDP

Catalog No. T158925G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
T158925G 25 g
T15895G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. T158925G Supplier TCI America Supplier No. T158925G
Add to Cart
Add to Cart

Chemical Identifiers

CAS 2105-61-5
Molecular Formula C6H2F3NO2
Molecular Weight (g/mol) 177.08
MDL Number MFCD00007051
InChI Key ROJNMGYMBLNTPK-UHFFFAOYSA-N
Synonym 2,4,5-trifluoronitrobenzene, benzene, 1,2,4-trifluoro-5-nitro, 1,2,5-trifluoro-4-nitrobenzene, 1-nitro-2,4,5-trifluorobenzene, 2,4,5-trifluoro-1-nitrobenzene, 2,4,5-trifluoro-nitrobenzene, 5-nitro-1,2,4-trifluorobenzene, 1,2,4-trifluoro-5-nitro-benzene, 1,2,4-trifluor-5-nitrobenzol
PubChem CID 75012
IUPAC Name 1,2,4-trifluoro-5-nitrobenzene
SMILES [O-][N+](=O)C1=CC(F)=C(F)C=C1F

Specifications

Boiling Point 195°C
Quantity 25 g
UN Number 2810
Formula Weight 177.08
Percent Purity ≥98.0% (GC)
Physical Form Liquid
Chemical Name or Material 2,4,5-Trifluoronitrobenzene
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.