Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

2,4,5-Trifluorophenylboronic acid, 97%, Thermo Scientific™

Catalog No. p-4380118
Click to view available options
Quantity:
5 g

Chemical Identifiers

CAS 247564-72-3
Molecular Formula C6H4BF3O2
Molecular Weight (g/mol) 175.90
MDL Number MFCD01863165
InChI Key KCHHKNCSISEAAE-UHFFFAOYSA-N
Synonym 2,4,5-trifluorophenyl boronic acid, 2,4,5-trifluorobenzeneboronic acid, boronic acid, b-2,4,5-trifluorophenyl, boronic acid, 2,4,5-trifluorophenyl, 2,4,5-trifluorophenylboronicacid, pubchem1787, acmc-1cd74
PubChem CID 2783133
IUPAC Name (2,4,5-trifluorophenyl)boronic acid
SMILES OB(O)C1=CC(F)=C(F)C=C1F

Specifications

Melting Point 118.0°C to 119.0°C
Assay Percent Range 96% min. (HPLC)
Packaging Glass bottle
Linear Formula F3C6H2B(OH)2
Quantity 5 g
Solubility Information Solubility in water: .. Other solubilities: soluble in methanol
Formula Weight 175.9
Percent Purity 97%
Chemical Name or Material 2, 4, 5-Trifluorophenylboronic acid
Hazard Category GHS Signal Word: Warning
Hazard Statement GHS H Statement
Harmful if swallowed.
Causes serious eye irritation.
May cause respiratory irritation.
Causes skin irritation.
Precautionary Statement GHS P Statement
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
IF ON SKIN: Wash

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.