Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

2,4,5-Trimethoxybenzoic Acid 98.0+%, TCI America™
SDP

Catalog No. T262725G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
T262725G 25 g
T26275G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. T262725G Supplier TCI America Supplier No. T262725G
Add to Cart
Add to Cart

Chemical Identifiers

CAS 490-64-2
Molecular Formula C10H11O5
Molecular Weight (g/mol) 211.19
MDL Number MFCD00002435
InChI Key KVZUCOGWKYOPID-UHFFFAOYSA-M
Synonym asaronic acid, benzoic acid, 2,4,5-trimethoxy, asarylic acid, calamonic acid, acmc-209ket, 2,4,5-trimethoxybenzoicacid, ksc489o7d, 2,4,5-trimethoxy-benzoic acid, 2,4,5-trimethoxybenzoic acid, alpha-hydroxyphenylacetic acid; amygdalic acid; alpha-hydroxyphenylacetic acid; alpha-hydroxy-benzeneacetic acid
PubChem CID 10276
IUPAC Name 2,4,5-trimethoxybenzoate
SMILES COC1=CC(OC)=C(C=C1OC)C([O-])=O

Specifications

Melting Point 144°C
Color White-Yellow
Boiling Point 300°C
Quantity 25 g
Formula Weight 212.20
Percent Purity ≥98.0% (GC,T)
Physical Form Crystalline Powder
Chemical Name or Material 2,4,5-Trimethoxybenzoic Acid
EINECSNumber (3)-4610
TSCA Yes
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.