Alfa Aesar™ 2,4,6-Collidine, 99%

Manufacturer: Alfa Aesar™ A11058AP

Quantity 500mL

 View more versions of this product

Catalog No. AAA11058AP

  Check Availability
Add to Cart

    Chemical Identifiers

    CAS 108-75-8
    Molecular Formula C8H11N
    Formula Weight 121.18
    MDL Number MFCD00006338
    UN Number UN1992
    Beilstein 107283
    Merck Index 14,9718
    Synonym 2,4,6-Trimethylpyridine
    Chemical Name or Material 2,4,6-Collidine

    Specifications

    Formula Weight 121.18
    Health Hazard 3 GHS P Statement
    P210-P303+P361+P353-P305+P351+P338-P361-P405-P501a
    Keep away from heat/sparks/open flames/hot surfaces.
    - No smoking.
    IF ON SKIN (or hair): Remove/Take off immediately all contaminated clothing. Rinse skin with water/shower.
    IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do.
    Continue rinsing.
    Remove/Take off immediately all contaminated clothing.
    Store locked up.
    Dispose of contents/container in accordance with local/regional/national/international regulations.
    TSCA TSCA
    Health Hazard 2 GHS H Statement
    H311-H226-H302-H315-H319-H335
    Toxic in contact with skin.
    Flammable liquid and vapor.
    Harmful if swallowed.
    Causes skin irritation.
    Causes serious eye irritation.
    May cause respiratory irritation.
    Quantity 500mL
    Odor Unpleasant
    MDL Number MFCD00006338
    CAS 108-75-8
    UN Number UN1992
    Synonym 2,4,6-Trimethylpyridine
    pH 9 to 10
    DOT Information Transport Hazard Class: 3; Packing Group: III; Proper Shipping Name: FLAMMABLE LIQUIDS, TOXIC, N.O.S.
    RTECS Number UU0970000
    Beilstein 107283
    Merck Index 14,9718
    Flash Point 54°C (129°F)
    Health Hazard 1 Danger
    Melting Point −43°C
    Boiling Point 171° to 172°C
    Molecular Formula C8H11N
    Chemical Name or Material 2,4,6-Collidine
    Assay 99%

    Description

    Applications
    2,4,6-Collidine is used as a tissue fixative for electron microscopy. It is useful in dehydrohalogenation reactions and acts as a solvent for the cleavage of hindered esters by anhydrous lithium iodide.

    Solubility
    Slightly miscible with water. Miscible with ether, methanol, acetone, chloroform, benzene, toluene and dilute acids.

    Notes
    Incompatible with oxidizing agents and strong acids.

    GHS H Statement
    H311-H226-H302-H315-H319-H335
    Toxic in contact with skin.
    Flammable liquid and vapor.
    Harmful if swallowed.
    Causes skin irritation.
    Causes serious eye irritation.
    May cause respiratory irritation.

    GHS P Statement
    P210-P303+P361+P353-P305+P351+P338-P361-P405-P501a
    Keep away from heat/sparks/open flames/hot surfaces.
    - No smoking.
    IF ON SKIN (or hair): Remove/Take off immediately all contaminated clothing. Rinse skin with water/shower.
    IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do.
    Continue rinsing.
    Remove/Take off immediately all contaminated clothing.
    Store locked up.
    Dispose of contents/container in accordance with local/regional/national/international regulations.

    Danger

    SDS & Safety Documents

  • Description & Specifications
  • Safety and Handling

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit