Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

2,4,6-Tribromophenol, 98%

Catalog No. aaa1242430
Click to view available options
Quantity:
250 g
1000 g
This item is not returnable. View return policy
This item is not returnable. View return policy

CAS: 118-79-6 | C6H3Br3O | 330.80 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 118-79-6
Molecular Formula C6H3Br3O
Molecular Weight (g/mol) 330.80
MDL Number MFCD00002150
InChI Key BSWWXRFVMJHFBN-UHFFFAOYSA-N
Synonym tribromophenol, bromol, bromkal pur 3, xeroform, phenol, 2,4,6-tribromo, flammex 3bp, unii-ys6k3eu393, ccris 1658, great lakes ph-73, 5175-83-7 bismuth 3+ salt
PubChem CID 1483
ChEBI CHEBI:47696
IUPAC Name 2,4,6-tribromophenol
SMILES OC1=C(Br)C=C(Br)C=C1Br

Specifications

Melting Point 86°C to 92°C
Density 2.55
Boiling Point 282°C to 290°C (sublimation)
Quantity 250 g
UN Number UN3077
Beilstein 776920
Merck Index 14,9613
Formula Weight 330.8
Percent Purity 98%
Chemical Name or Material 2,4,6-Tribromophenol
Hazard Category H317-H319
Hazard Statement GHS H Statement
H301-H332-H315-H319-H335
Toxic if swallowed.
Harmful if inhaled.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P272-P280-P302+P352-P305+P351+P338-P333+P313-P363-P501c
DOTInformation Transport Hazard Class: 9; Packing Group: III; Proper Shipping Name: ENVIRONMENTALLY HAZARDOUS SUBSTANCES, SOLID, N.O.S.
EINECSNumber 204-278-6
RTECSNumber SN1225000
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.