Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

2,4,6-Trichlorophenol, 98%

Catalog No. p-7081629
Click to view available options
Quantity:
250 g
1000 g

CAS: 88-06-2 | C6H3Cl3O | 197.439 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Suitable for battery materials development.

Applications
2,4,6-Trichlorophenol, antiseptic, defoliant, and glue preservative, chemical intermediate, anti-mildew treatment, solvent for polyester fiber.

Solubility
Soluble in water(0.8g/L).

Notes
Keep container tightly closed. Store in cool, dry conditions in well sealed containers.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 88-06-2
Molecular Formula C6H3Cl3O
MDL Number MFCD00002172
InChI Key LINPIYWFGCPVIE-UHFFFAOYSA-N
PubChem CID 6914
ChEBI CHEBI:28755
IUPAC Name 2,4,6-trichlorophenol

Specifications

Melting Point 63°C to 68°C
Density 1.675
Boiling Point 245°C to 246°C
Flash Point 99°C (210°F)
Assay Percent Range 98%
Quantity 250 g
UN Number UN2020
Beilstein 776729
Merck Index 14,9644
Synonym dowicide 2s, phenachlor, phenol, 2,4,6-trichloro, omal, phenaclor, rcra waste number u231, dowcide 2s, 1,3,5-trichloro-2-hydroxybenzene, 2,4,6-trichlorfenol, unii-mhs8c5bauz
Solubility Information Soluble in water(0.8g/L).
SMILES C1=C(C=C(C(=C1Cl)O)Cl)Cl
Molecular Weight (g/mol) 197.439
Formula Weight 197.45
Percent Purity 98%
Chemical Name or Material 2,4,6-Trichlorophenol, 98%
Show More Show Less
Hazard Category H302-H315-H319-H351
Hazard Statement GHS H Statement
H351-H302-H315-H319
Precautionary Statement P201-P202-P264b-P270-P280i-P281-P301+P312-P302+P352-P305+P351+P338-P308+P313-P330-P332+P313-P362-P501c
DOTInformation Transport Hazard Class: 6.1; Packing Group: III; Proper Shipping Name: CHLOROPHENOLS, SOLID
EINECSNumber 201-795-9
RTECSNumber SN1575000
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.