Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

2,4,6-Trichlorophenol, 98%

Catalog No. AC139485000
Click to view available options
Quantity:
100 g
500 g
Packaging:
Glass bottle

Catalog No. AC139485000

Supplier: Thermo Scientific Chemicals 139485000

Only null left
Add to Cart

CAS: 88-06-2 | C6H3Cl3O | 197.45 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 88-06-2
Molecular Formula C6H3Cl3O
Molecular Weight (g/mol) 197.45
MDL Number MFCD00002172
InChI Key LINPIYWFGCPVIE-UHFFFAOYSA-N
Synonym dowicide 2s, phenachlor, phenol, 2,4,6-trichloro, omal, phenaclor, rcra waste number u231, dowcide 2s, 1,3,5-trichloro-2-hydroxybenzene, 2,4,6-trichlorfenol, unii-mhs8c5bauz
PubChem CID 6914
ChEBI CHEBI:28755
IUPAC Name 2,4,6-trichlorophenol
SMILES C1=C(C=C(C(=C1Cl)O)Cl)Cl

Specifications

CAS Min % 97.5
CAS Max % 100.0
Melting Point 64.0°C to 68.0°C
Color Beige to Brown
Boiling Point 246.0°C
Flash Point 99°C
Infrared Spectrum Authentic
Assay Percent Range 97.5% min. (GC)
Packaging Glass bottle
Linear Formula Cl3C6H2OH
Quantity 500 g
Beilstein 06, 190
Merck Index 15, 9809
Solubility Information Solubility in water: 0.8g/L. Other solubilities: soluble in aromatic solvents and caustic soda sol.,soluble in (g/100g) acetone 525; benzene 113;,ccl4 37; diacetone alcohol 335; ether 354;,methanol 525; pine oil 163; toluene 100;,turpentine 37
Formula Weight 197.45
Percent Purity 98%
Physical Form Crystalline Mass or Crystalline Powder and Chunks
Chemical Name or Material 2, 4, 6-Trichlorophenol
Show More Show Less
Hazard Category GHS Signal Word: Warning
Hazard Statement GHS H Statement
Causes skin irritation.
Causes serious eye irritation.
Suspected of causing cancer.
Harmful if swallowed.
Very toxic to aquatic life with long lasting effects.
Precautionary Statement GHS P Statement
Wear protective gloves/protective clothing/eye protection/face protection.
IF ON SKIN: Gently wash with plenty of soap and water.
IF SWALLOWED: rinse mouth.
Do NOT induce vomiting.
Call a POISON CENTER or doctor/p
EINECSNumber 201-795-9

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.