Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

2,4,6-Trichlorophenoxyacetic Acid 98.0+%, TCI America™
SDP

Catalog No. T106325G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
T106325G 25 g
T10635G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. T106325G Supplier TCI America Supplier No. T106325G
Add to Cart
Add to Cart

Chemical Identifiers

CAS 575-89-3
Molecular Formula C8H5Cl3O3
Molecular Weight (g/mol) 255.475
MDL Number MFCD00053498
InChI Key KZDCLQBOHGBWOI-UHFFFAOYSA-N
PubChem CID 11331
IUPAC Name 2-(2,4,6-trichlorophenoxy)acetic acid
SMILES C1=C(C=C(C(=C1Cl)OCC(=O)O)Cl)Cl

Specifications

Melting Point 185°C
Color White
Quantity 25 g
Physical Form Crystal-Powder at 20°C
UN Number 2811
Formula Weight 255.48
Percent Purity ≥98.0% (GC,T)
Chemical Name or Material 2,4,6-Trichlorophenoxyacetic Acid
RTECSNumber AJ8405000
TSCA No
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.