Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

2,4,6-Trifluoronitrobenzene 97.0+%, TCI America™
SDP

Catalog No. T168125G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
T168125G 25 g
T16815G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. T168125G Supplier TCI America Supplier No. T168125G
Add to Cart
Add to Cart

Chemical Identifiers

CAS 315-14-0
Molecular Formula C6H2F3NO2
Molecular Weight (g/mol) 177.082
MDL Number MFCD00014687
InChI Key PWRFDGYYJWQIAB-UHFFFAOYSA-N
Synonym 2,4,6-trifluoronitrobenzene, 2,2,6-trifluoronitrobenzene, benzene, 1,3,5-trifluoro-2-nitro, 1,3,5-trifluoro-2-nitro-benzene, 2-nitro-1,3,5-trifluorobenzene, pubchem10646, acmc-209hn4, 1,3,5-trifluoronitrobenzene, 2,4,6 trifluoronitrobenzene, ksc226o4t
PubChem CID 67567
IUPAC Name 1,3,5-trifluoro-2-nitrobenzene
SMILES C1=C(C=C(C(=C1F)[N+](=O)[O-])F)F

Specifications

Color Yellow
Boiling Point 172°C
Quantity 25 g
Formula Weight 177.08
Percent Purity ≥97.0% (GC)
Physical Form Liquid
Chemical Name or Material 2,4,6-Trifluoronitrobenzene
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.