Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

2,4,6-Trifluorophenylboronic Acid (contains varying amounts of Anhydride), TCI America™
SDP

Catalog No. p-7193800 Shop All Tokyo Chemical Industry (TCI) Products
Click to view available options
Quantity:
1 g
5 g

Specifications

Chemical Name or Material 2,4,6-Trifluorophenylboronic Acid (contains varying amounts of Anhydride)
CAS 182482-25-3
Melting Point 235°C
Molecular Formula C6H4BF3O2
Quantity 1 g
MDL Number MFCD01863169
Synonym 2,4,6-trifluorophenyl boronic acid, 2,4,6-trifluorobenzeneboronic acid, 2,4,6-trifluorophenylboronicacid, 2,4,6-trifluorophenyl boranediol, boronic acid, 2,4,6-trifluorophenyl, pubchem1788, acmc-209ejj, chembl91770, boronicacid, b-2,4,6-trifluorophenyl, boronic acid, b-2,4,6-trifluorophenyl
InChI Key IPEIGKHHSZFAEW-UHFFFAOYSA-N
SMILES OB(O)C1=C(F)C=C(F)C=C1F
IUPAC Name (2,4,6-trifluorophenyl)boronic acid
Molecular Weight (g/mol) 175.90
PubChem CID 2779329
Formula Weight 175.90
Physical Form Crystalline Powder
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.