Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

2',4',6'-Trihydroxy-2-phenylacetophenone 96.0+%, TCI America™
SDP

Catalog No. T33901G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
T33901G 1 g
T33905G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. T33901G Supplier TCI America Supplier No. T33901G
Add to Cart
Add to Cart

Chemical Identifiers

CAS 727-71-9
Molecular Formula C14H12O4
Molecular Weight (g/mol) 244.25
MDL Number MFCD00205539
InChI Key SLHBRIIHMDJIBT-UHFFFAOYSA-N
Synonym Benzyl 2,4,6-Trihydroxyphenyl Ketone
PubChem CID 689111
IUPAC Name 2-phenyl-1-(2,4,6-trihydroxyphenyl)ethan-1-one
SMILES OC1=CC(O)=C(C(=O)CC2=CC=CC=C2)C(O)=C1

Specifications

Melting Point 166°C
Color Yellow
Quantity 1 g
UN Number 3077
Formula Weight 244.25
Percent Purity ≥96.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material 2′,4′,6′-Trihydroxy-2-phenylacetophenone
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.