Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

2,4,6-Triisopropylbenzoyl chloride, 98+%

Catalog No. p-7081342
Click to view available options
Quantity:
5 g
25 g

CAS: 57199-00-5 | C16H23ClO | 266.809 g/mol

2,4,6-Triisopropylbenzoyl chloride is used as pharmaceuticcal intermediates.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
2,4,6-Triisopropylbenzoyl chloride is used as pharmaceuticcal intermediates.

Solubility
Insoluble in water.

Notes
Moisture Sensitive. Store away from moisture and oxidizing agents. Incompatible with strong oxidizing agents, ammonia etc,.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 57199-00-5
Molecular Formula C16H23ClO
Molecular Weight (g/mol) 266.809
MDL Number MFCD00015030
InChI Key OSKNTKJPGKHDHV-UHFFFAOYSA-N
Synonym 2,4,6-triisopropylbenzoyl chloride, benzoyl chloride, 2,4,6-tris 1-methylethyl, acmc-20aoev, 2,4,6-triiso-propylbenzoyl chloride, 2,4,6-tri propan-2-yl benzoyl chloride, benzoyl chloride,2,4,6-tris 1-methylethyl, 2,4,6-tris 1-methylethyl benzoic acid chloride
PubChem CID 92697
IUPAC Name 2,4,6-tri(propan-2-yl)benzoyl chloride
SMILES CC(C)C1=CC(=C(C(=C1)C(C)C)C(=O)Cl)C(C)C

Specifications

Melting Point 78°C to 82°C
Quantity 5 g
UN Number UN3261
Beilstein 2269534
Sensitivity Moisture sensitive
Solubility Information Insoluble in water.
Formula Weight 266.81
Percent Purity ≥98%
Chemical Name or Material 2,4,6-Triisopropylbenzoyl chloride
Hazard Category H314-H335
Hazard Statement GHS H Statement
H314-H318
Causes severe skin burns and eye damage.
Causes serious eye damage.
Precautionary Statement P260-P264b-P271-P280-P301+P330+P331-P303+P361+P353-P304+P340-P305+P351+P338-P310-P363-P501c
DOTInformation Transport Hazard Class: 8; Packing Group: II; Proper Shipping Name: CORROSIVE SOLID, ACIDIC, ORGANIC, N.O.S.
EINECSNumber 260-614-1
TSCA No
Recommended Storage Ambient temperatures; Store under Nitrogen

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.