Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

2,4,6-Trimethoxybenzyl alcohol, 97%

Catalog No. p-7033270
Click to view available options
Quantity:
250 mg
1 g

CAS: 61040-78-6 | C10H14O4 | 198.218 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 61040-78-6
Molecular Formula C10H14O4
Molecular Weight (g/mol) 198.218
MDL Number MFCD00060356
InChI Key CFXXBVNHYJQNKS-UHFFFAOYSA-N
PubChem CID 602515
IUPAC Name (2,4,6-trimethoxyphenyl)methanol
SMILES COC1=CC(=C(C(=C1)OC)CO)OC

Specifications

Melting Point 60°C
Quantity 250 mg
Synonym 2,4,6-trimethoxyphenyl methanol, 2,4,6-trimethoxybenzyl alcohol, 2,4,6-trimethoxyphenyl methan-1-ol, 2,4,6-trimethoxy-benzyl alcohol, benzenemethanol,2,4,6-trimethoxy, benzenemethanol, 2,4,6-trimethoxy, 2,4,6-trimethoxy-phenyl-methanol, 2,4,6-trimethoxyphenyl methanol #, 3-amino-n-1,1-dimethylethoxy carbonyl-l-alanine methyl ester
Formula Weight 198.22
Percent Purity 97%
Chemical Name or Material 2,4,6-Trimethoxybenzyl alcohol
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.