Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

2,4,6-Trimethyl-2,4,6-trivinylcyclotrisilazane 97.0+%, TCI America™
SDP

Catalog No. T255625G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
T255625G 25 g
T25565G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. T255625G Supplier TCI America Supplier No. T255625G
Add to Cart
Add to Cart

Chemical Identifiers

CAS 5505-72-6
Molecular Formula C9H21N3Si3
Molecular Weight (g/mol) 255.543
MDL Number MFCD00054934
InChI Key RFSBGZWBVNPVNN-UHFFFAOYSA-N
PubChem CID 79644
IUPAC Name 2,4,6-tris(ethenyl)-2,4,6-trimethyl-1,3,5,2,4,6-triazatrisilinane
SMILES C[Si]1(N[Si](N[Si](N1)(C)C=C)(C)C=C)C=C

Specifications

Color Yellow
Boiling Point 65°C
Quantity 25 g
UN Number 3267
Formula Weight 255.54
Percent Purity ≥97.0% (T)
Physical Form Liquid
Chemical Name or Material 2,4,6-Trimethyl-2,4,6-trivinylcyclotrisilazane
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.