Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

2',4',6'-Trimethylacetophenone, 97+%

Catalog No. p-7081535
Click to view available options
Quantity:
25 g
100 g

CAS: 1667-01-2 | C11H14O | 162.23 g/mol

2',4',6'-Trimethylacetophenone is used as laboratory chemicals, manufacture of substances and intermediated for chemical synthesis.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
2′,4′,6′-Trimethylacetophenone is used as laboratory chemicals, manufacture of substances and intermediated for chemical synthesis.

Solubility
Difficult to mix.

Notes
Store in cool, dry conditions in well sealed containers. Keep container tightly closed.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 1667-01-2
Molecular Formula C11H14O
Molecular Weight (g/mol) 162.23
MDL Number MFCD00008735
InChI Key XWCIICLTKWRWCI-UHFFFAOYSA-N
Synonym 1-mesitylethanone, 2',4',6'-trimethylacetophenone, acetylmesitylene, acetomesitylene, mesityl methyl ketone, 1-2,4,6-trimethylphenyl ethanone, ethanone, 1-2,4,6-trimethylphenyl, 2-acetylmesitylene, 2,4,6-trimethylacetophenone, methyl 2,4,6-trimethylphenyl ketone
PubChem CID 15461
IUPAC Name 1-(2,4,6-trimethylphenyl)ethanone
SMILES CC(=O)C1=C(C)C=C(C)C=C1C

Specifications

Density 0.975
Boiling Point 235°C to 236°C
Flash Point >110°C (230°F)
Refractive Index 1.517
Quantity 25 g
Beilstein 2044595
Solubility Information Difficult to mix.
Formula Weight 162.23
Percent Purity 98%
Chemical Name or Material 2',4',6'-Trimethylacetophenone
EINECSNumber 216-783-9
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.