Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

2,4,6-Trimethylaniline 99.0+%, TCI America™
SDP

Catalog No. T0467100ML Shop All Tokyo Chemical Industry (TCI) Products
Click to view available options
Quantity:
25 mL
100 mL
500 mL

Catalog No. T0467100ML

Supplier: TCI America T0467100ML

Only null left
Add to Cart
Add to Cart

Chemical Identifiers

CAS 88-05-1
Molecular Formula C9H13N
Molecular Weight (g/mol) 135.21
MDL Number MFCD00007740
InChI Key KWVPRPSXBZNOHS-UHFFFAOYSA-N
Synonym mesidine, aminomesitylene, mesitylamine, benzenamine, 2,4,6-trimethyl, mesidin, 2,4,6-trimethylbenzenamine, 2-aminomesitylene, 2-amino-1,3,5-trimethylbenzene, 2,4,6-trimethyl aniline, mesitylene, 2-amino
PubChem CID 6913
ChEBI CHEBI:82545
IUPAC Name 2,4,6-trimethylaniline
SMILES CC1=CC(=C(C(=C1)C)N)C

Specifications

Boiling Point 230°C
Quantity 100 mL
UN Number 2810
Formula Weight 135.21
Percent Purity ≥99.0% (GC)
Physical Form Liquid
Chemical Name or Material 2,4,6-Trimethylaniline
EINECSNumber (3)-0132
RTECSNumber BZ0700000
TSCA Yes
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.