Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Alfa Aesar™ 2,4,6-Trimethylbenzeneboronic acid, 97%

Click to view available options
Quantity:
1g
5g
25g
5980-97-2
C9H13BO2
164.011
MFCD00236060
BZXQRXJJJUZZAJ-UHFFFAOYSA-N
mesitylboronic acid,2,4,6-trimethylbenzeneboronic acid,2-mesityleneboronic acid,2,4,6-trimethylphenyl boronic acid,2,4,6-trimethylphenyl boranediol,boronic acid, 2,4,6-trimethylphenyl,2,4,6-trimethylboronic acid,boronic acid, b-2,4,6-trimethylphenyl,mesityl boronic acid,pubchem7895
292184
(2,4,6-trimethylphenyl)boronic acid
B(C1=C(C=C(C=C1C)C)C)(O)O
5980-97-2
C9H13BO2
164.011
MFCD00236060
BZXQRXJJJUZZAJ-UHFFFAOYSA-N
mesitylboronic acid,2,4,6-trimethylbenzeneboronic acid,2-mesityleneboronic acid,2,4,6-trimethylphenyl boronic acid,2,4,6-trimethylphenyl boranediol,boronic acid, 2,4,6-trimethylphenyl,2,4,6-trimethylboronic acid,boronic acid, b-2,4,6-trimethylphenyl,mesityl boronic acid,pubchem7895
292184
(2,4,6-trimethylphenyl)boronic acid
B(C1=C(C=C(C=C1C)C)C)(O)O
Quantity 1g
Melting Point ~120°C to 127°C
Beilstein 2831927
Formula Weight 164.01
Percent Purity 97%
Chemical Name or Material 2,4,6-Trimethylbenzeneboronic acid

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Warning