Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

2,4,6-Trimethylbenzenesulfonyl hydrazide, 97%, Thermo Scientific™

Catalog No. p-206509
Change view
Click to view available options
Quantity:
5 g
Packaging:
Glass Bottle
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
AC185090050 5 g Glass Bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AC185090050 Supplier Thermo Scientific Chemicals Supplier No. 185090050

Chemical Identifiers

CAS 16182-15-3
MDL Number MFCD00007585
InChI Key JQUBKTQDNVZHIY-UHFFFAOYSA-N
Synonym 2,4,6-trimethylbenzenesulfonyl hydrazide, mesitylene-2-sulphonohydrazide, 2-mesitylenesulfonyl hydrazide, 2-mesitylenesulfonohydrazide, 2-mesitylenesulfono hydrazide, benzenesulfonic acid,2,4,6-trimethyl-, hydrazide, hydrazino 2,4,6-trimethylphenyl sulfone, mesitylenesulphonohydrazide, mesitylenesulfonylhydrazide, 2-mesitylehesulfonohydrazide
PubChem CID 85321
IUPAC Name 2,4,6-trimethylbenzenesulfonohydrazide
SMILES CC1=CC(=C(C(=C1)C)S(=O)(=O)NN)C

Specifications

CAS Min % 96.0
CAS Max % 100.0
Melting Point 110.0°C to 114.0°C
Color White
Infrared Spectrum Authentic
Assay Percent Range 97%
Packaging Glass Bottle
Molecular Formula C9H14N2O2S
Linear Formula (CH3)3C6H2SO2NHNH2
Quantity 5 g
Molecular Weight (g/mol) 214.283
Formula Weight 214.28
Percent Purity 97%
Physical Form Crystalline Powder
Chemical Name or Material 2, 4, 6-Trimethylbenzenesulfonyl hydrazide
Show More Show Less
Hazard Category Warning
Hazard Statement GHS H Statement:
Causes serious eye irritation.
May cause respiratory irritation.
Causes skin irritation.
Flammable solid.
Precautionary Statement GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
EINECSNumber 240-319-4
TSCA TSCA

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.