Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

2,4,6-Trimethylbenzoic acid, 99%

Catalog No. p-7081735
Click to view available options
Quantity:
10 g
50 g
250 g

CAS: 480-63-7 | C10H12O2 | 164.20 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 480-63-7
Molecular Formula C10H12O2
Molecular Weight (g/mol) 164.20
MDL Number MFCD00002481
InChI Key FFFIRKXTFQCCKJ-UHFFFAOYSA-N
Synonym mesitoic acid, mesitylenecarboxylic acid, beta-isodurylic acid, benzoic acid, 2,4,6-trimethyl, .beta.-isodurylic acid, unii-4pu2hg6787, 2-mesitylenecarboxylic acid, 2,4,6-trimethyl-benzoic acid, 2,6-trimethylbenzoic acid, benzoic acid,4,6-trimethyl
PubChem CID 10194
ChEBI CHEBI:64830
IUPAC Name 2,4,6-trimethylbenzoic acid
SMILES CC1=CC(C)=C(C(O)=O)C(C)=C1

Specifications

Melting Point 153°C to 155°C
Quantity 10 g
Beilstein 1866187
Formula Weight 164.2
Percent Purity 99%
Chemical Name or Material 2,4,6-Trimethylbenzoic acid
EINECSNumber 207-553-9
RTECSNumber DI0887010
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.