Alfa Aesar™ 2,4,6-Trimethylbenzonitrile, 98%

Manufacturer: Alfa Aesar™ A1188214

Quantity 25g

 View more versions of this product

Catalog No. AAA1188214

  Check Availability
Add to Cart

    Chemical Identifiers

    CAS 2571-52-0
    Molecular Formula C10H11N
    Formula Weight 145.21
    MDL Number MFCD00016378
    UN Number UN3439
    Beilstein 2044618
    Synonym Cyanomesitylene; Mesitonitrile
    Chemical Name or Material 2,4,6-Trimethylbenzonitrile

    Specifications

    Formula Weight 145.21
    Health Hazard 3 GHS P Statement
    P261-P280-P305+P351+P338-P304+P340-P405-P501a
    Avoid breathing dust/fume/gas/mist/vapors/spray.
    Wear protective gloves/protective clothing/eye protection/face protection.
    IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do.
    Continue rinsing.
    IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
    Store locked up.
    Dispose of contents/container in accordance with local/regional/national/international regulations.
    MDL Number MFCD00016378
    Molecular Formula C10H11N
    CAS 2571-52-0
    Beilstein 2044618
    Synonym Cyanomesitylene; Mesitonitrile
    Health Hazard 1 Danger
    DOT Information Transport Hazard Class: 6.1; Packing Group: III; Proper Shipping Name: NITRILES, SOLID, TOXIC, N.O.S.
    Boiling Point 79° to 80°C (2mmHg)
    Melting Point 50° to 55°C
    Quantity 25g
    UN Number UN3439
    Health Hazard 2 GHS H Statement
    H331-H302-H312-H315-H319-H335
    Toxic if inhaled.
    Harmful if swallowed.
    Harmful in contact with skin.
    Causes skin irritation.
    Causes serious eye irritation.
    May cause respiratory irritation.
    Chemical Name or Material 2,4,6-Trimethylbenzonitrile
    Assay 98%

    Description

    Applications
    It employed as a intermediate for pharmaceutical.

    Solubility
    Slightly soluble in water.

    Notes
    Store in cool, dry conditions in well sealed container. Store away from oxidizing agent.

    GHS H Statement
    H331-H302-H312-H315-H319-H335
    Toxic if inhaled.
    Harmful if swallowed.
    Harmful in contact with skin.
    Causes skin irritation.
    Causes serious eye irritation.
    May cause respiratory irritation.

    GHS P Statement
    P261-P280-P305+P351+P338-P304+P340-P405-P501a
    Avoid breathing dust/fume/gas/mist/vapors/spray.
    Wear protective gloves/protective clothing/eye protection/face protection.
    IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do.
    Continue rinsing.
    IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
    Store locked up.
    Dispose of contents/container in accordance with local/regional/national/international regulations.

    Danger

    SDS & Safety Documents

  • Description & Specifications
  • Safety and Handling

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit