Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

2,4,6-Trimethylphenol, 99%

Catalog No. AC140210500
Click to view available options
Quantity:
10 g
50 g
Packaging:
Glass bottle

CAS: 527-60-6 | C9H12O | 136.19 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 527-60-6
Molecular Formula C9H12O
Molecular Weight (g/mol) 136.19
MDL Number MFCD00002235
InChI Key BPRYUXCVCCNUFE-UHFFFAOYSA-N
Synonym mesitol, phenol, 2,4,6-trimethyl, mesityl alcohol, 2-hydroxymesitylene, 2,4,6-trimetylofenol, 1,3,5-trimethylphenol, 2,4,6-trimethyl-phenol, 1-hydroxy-2,4,6-trimethylbenzene, 2,4,6-trimethylofenol, unii-fpz32614n6
PubChem CID 10698
IUPAC Name 2,4,6-trimethylphenol
SMILES CC1=CC(=C(C(=C1)C)O)C

Specifications

CAS Min % 98.5
CAS Max % 100.0
Melting Point 70°C to 73°C
Color Beige
Boiling Point 220°C
Infrared Spectrum Authentic
Assay Percent Range 98.5% min. (GC)
Packaging Glass bottle
Linear Formula (CH3)3C6H2OH
Quantity 50 g
Beilstein 06, 518
Formula Weight 136.19
Percent Purity 99%
Physical Form Crystalline Needles
Chemical Name or Material 2, 4, 6-Trimethylphenol
Show More Show Less
Hazard Category GHS Signal Word: Danger
Hazard Statement GHS H Statement
Harmful if swallowed.
Harmful in contact with skin.
Causes severe skin burns and eye damage.
May cause an allergic skin reaction.
May cause respiratory irritation.
May cause damage to organs through pr
Precautionary Statement GHS P Statement
Wear protective gloves/protective clothing/eye protection/face protection.
IF SWALLOWED: Rinse mouth.
Do NOT induce vomiting.
IF ON SKIN (or hair): Take off immediately all contaminated clothing.
Rinse skin with wat
EINECSNumber 208-419-2

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.