Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

2,4,6-Trimethylphenylboronic acid, 95%

Catalog No. AC359790010
Click to view available options
Quantity:
1 g

CAS: 5980-97-2 | C9H13BO2 | 164.01 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 5980-97-2
Molecular Formula C9H13BO2
Molecular Weight (g/mol) 164.01
InChI Key BZXQRXJJJUZZAJ-UHFFFAOYSA-N
Synonym mesitylboronic acid, 2,4,6-trimethylbenzeneboronic acid, 2-mesityleneboronic acid, 2,4,6-trimethylphenyl boronic acid, 2,4,6-trimethylphenyl boranediol, boronic acid, 2,4,6-trimethylphenyl, 2,4,6-trimethylboronic acid, boronic acid, b-2,4,6-trimethylphenyl, mesityl boronic acid, pubchem7895
PubChem CID 292184
IUPAC Name (2,4,6-trimethylphenyl)boronic acid
SMILES B(C1=C(C=C(C=C1C)C)C)(O)O

Specifications

Melting Point 131.0°C to 136.0°C
Infrared Spectrum Authentic
Assay Percent Range 94% min. (HPLC)
Packaging Glass Bottle
Quantity 1 g
Formula Weight 164.01
Percent Purity 95%
Chemical Name or Material 2,4,6-Trimethylphenylboronic acid
Hazard Category GHS Signal Word: Warning
Hazard Statement GHS H Statement
Causes skin irritation.
Causes serious eye irritation.
Precautionary Statement GHS P Statement
Wear protective gloves/protective clothing/eye protection/face protection.
IF ON SKIN: Wash with plenty of soap and water.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses,if present a

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.