Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Tris(dibenzylideneacetone)dipalladium-chloroform adduct, 97%

Catalog No. AC369340010
Click to view available options
Quantity:
1 g
5 g
Packaging:
Glass bottle

Catalog No. AC369340010

Supplier: Thermo Scientific Chemicals 369340010

Only null left
Add to Cart

CAS: 52522-40-4 | C52H43Cl3O3Pd2 | 1035.10 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 52522-40-4
Molecular Formula C52H43Cl3O3Pd2
Molecular Weight (g/mol) 1035.10
MDL Number MFCD00075479
InChI Key LNAMMBFJMYMQTO-UHFFFAOYSA-N
Synonym tris dibenzylideneacetone dipalladium-chloroform adduct, tris dibenylideneacetone dipalladium-chloroform, tris dibenzylideneacetone dipalladium 0 chloroform adduct, tris dibenzylideneacetone dipalladium 0-chloroform adduct, tris dibenzylideneacetone dipalladium chloroform adduct, tris dibenzylideneacetone dipalladium-chloroform, pd2 dba 3 chcl3, tris dibenzylideneacetone chloroform-di-palladium 0, tris dibenzylideneacetone dipalladium chloroform complex
PubChem CID 11029508
IUPAC Name chloroform;(1E,4E)-1,5-diphenylpenta-1,4-dien-3-one;palladium
SMILES [Pd].[Pd].ClC(Cl)Cl.O=C(C=CC1=CC=CC=C1)C=CC1=CC=CC=C1.O=C(C=CC1=CC=CC=C1)C=CC1=CC=CC=C1.O=C(C=CC1=CC=CC=C1)C=CC1=CC=CC=C1

Specifications

Melting Point 131.0°C to 135.0°C
Color Black to Purple
Assay Percent Range 96% min. (ex Pd)
Packaging Glass bottle
Linear Formula (C6H5CH=CHCOCH=CHC6H5)3Pd2·CHCl3
Quantity 1 g
Fieser 14,339; 15,353; 16,372; 17,394
Formula Weight 1035.11
Percent Purity 97%
Physical Form Crystalline Powder
Chemical Name or Material Tris(dibenzylideneacetone)dipalladium-chloroform adduct
Show More Show Less
Hazard Category GHS Signal Word: Warning
Hazard Statement GHS H Statement
Causes skin irritation.
Harmful if swallowed.
Suspected of causing cancer.
Precautionary Statement GHS P Statement
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.