Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

2,4,6-Trimethylthiophenol, 97%

Catalog No. p-7027781
Click to view available options
Quantity:
1 g
5 g
25 g

Catalog No. AAB2479803

Supplier: Thermo Scientific Chemicals B2479803

Only null left
Add to Cart

CAS: 1541-10-2 | C9H12S | 152.255 g/mol

2,4,6-Trimethylthiophenol, is used as pharmaceutical intermediate.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
2,4,6-Trimethylthiophenol, is used as pharmaceutical intermediate.

Solubility
Soluble in alcohol. Insoluble in water.

Notes
Store away from strong oxidizing agents. Keep container tightly closed. Store in cool, dry conditions in well sealed containers.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 1541-10-2
Molecular Formula C9H12S
Molecular Weight (g/mol) 152.255
MDL Number MFCD00218475
InChI Key PKANQZUPJCMBAK-UHFFFAOYSA-N
Synonym 2,4,6-trimethylthiophenol, 2,4,6-trimethylbenzene-1-thiol, benzenethiol,2,4,6-trimethyl, 2-mercaptomesitylene, 2-sulphanylmesitylene, acmc-20aoem, 2,4,6-trimethyl-benzene-1-thiol
PubChem CID 284449
IUPAC Name 2,4,6-trimethylbenzenethiol
SMILES CC1=CC(=C(C(=C1)C)S)C

Specifications

Quantity 1 g
Sensitivity Air Sensitive
Solubility Information Soluble in alcohol. Insoluble in water.
Formula Weight 152.25
Percent Purity 97%
Chemical Name or Material 2,4,6-Trimethylthiophenol
Hazard Category H315-H319-H335
Hazard Statement GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.