Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

2-(4-Bromophenyl)-1,1-diphenylethylene 98.0+%, TCI America™
SDP

Catalog No. B309025G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
B309025G 25 g
B30905G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. B309025G Supplier TCI America Supplier No. B309025G

Chemical Identifiers

CAS 18648-66-3
Molecular Formula C20H15Br
Molecular Weight (g/mol) 335.24
MDL Number MFCD01790865
InChI Key HUCFDUOIMSRYAA-UHFFFAOYSA-N
PubChem CID 630600
IUPAC Name 1-bromo-4-(2,2-diphenylethenyl)benzene
SMILES BrC1=CC=C(C=C(C2=CC=CC=C2)C2=CC=CC=C2)C=C1

Specifications

Melting Point 79°C
Color White-Yellow
Quantity 25 g
Formula Weight 335.24
Percent Purity ≥98.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material 2-(4-Bromophenyl)-1,1-diphenylethylene
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.